Learn More
Eurofins Discoverx PathHunter U2OS PD-L1/PD-L1 Dimerization Cell Line, 2 vials. (93-1129C3)
Catalog No. 502002151
Encompass
The PathHunter PD-L1/PD-L1 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. anti PD-L1 antibody) induced dimerization of PD-L1/PD-L1 receptors. This product uses EFC technology. The cell line included overexpresses PD-L1 target protein fused to PK alongside PD-L1 target protein fused to EA. Upon ligand-induced dimerization of PD-L1/PD-L1, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.
Catalog No. 50-200-2151 Supplier Eurofins Discoverx Supplier No. 931129C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter PD-L1/PD-L1 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. anti PD-L1 antibody) induced dimerization of PD-L1/PD-L1 receptors. This product uses EFC technology. The cell line included overexpresses PD-L1 target protein fused to PK alongside PD-L1 target protein fused to EA. Upon ligand-induced dimerization of PD-L1/PD-L1, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.