Learn More
Eurofins Discoverx PathHunter U2OS PTGER1 β-Arrestin Cell Line, 2 vials. (93-0937C3)
Catalog No. 502001609
Encompass
The PathHunter PTGER1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin E2) based activation of PTGER1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTGER1 and EA-tagged β-Arrestin-2. Activation of PTGER1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1609 Supplier Eurofins Discoverx Supplier No. 930937C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter PTGER1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin E2) based activation of PTGER1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged PTGER1 and EA-tagged β-Arrestin-2. Activation of PTGER1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.