Learn More
Eurofins Discoverx PathHunter U2OS RANK-IκB Functional Assay, 2 vials. (93-0994C3)
Catalog No. 502001769
Encompass
The PathHunter U2OS RANK-IκB Functional Assay is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting the NFκB pathway. This cell line enables detection of IκB levels, allowing for assessment of NFκB pathway activation-induced IκB degradation. This product uses EFC technology. The cell line included overexpresses IκB target protein fused to ProLabel alongside untagged RANK. EA is contained in the detection reagent. Upon pathway-activation-induced degradation of IκB EFC signal is lost. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.
Catalog No. 50-200-1769 Supplier Eurofins Discoverx Supplier No. 930994C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter U2OS RANK-IκB Functional Assay is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting the NFκB pathway. This cell line enables detection of IκB levels, allowing for assessment of NFκB pathway activation-induced IκB degradation. This product uses EFC technology. The cell line included overexpresses IκB target protein fused to ProLabel alongside untagged RANK. EA is contained in the detection reagent. Upon pathway-activation-induced degradation of IκB EFC signal is lost. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.