Learn More
Eurofins Discoverx PathHunter U2OS rFFAR1 β-Arrestin Cell Line, 2 vials. (93-0976C3)
Supplier:  Eurofins Discoverx 930976C3
Encompass
The PathHunter rFFAR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of rFFAR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rFFAR1 and EA-tagged β-Arrestin-2. Activation of rFFAR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1726
Add to Cart
Request a Quote