Learn More
Eurofins Discoverx PathHunter U2OS rHTR2C β-Arrestin Cell Line, 2 vials. (93-0915C3)
Catalog No. 502001556
Encompass
The PathHunter rHTR2C cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of rHTR2C GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rHTR2C and EA-tagged β-Arrestin-2. Activation of rHTR2C stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1556 Supplier Eurofins Discoverx Supplier No. 930915C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter rHTR2C cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin) based activation of rHTR2C GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged rHTR2C and EA-tagged β-Arrestin-2. Activation of rHTR2C stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Rat version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.