Learn More
Eurofins Discoverx PathHunter U2OS rhTrkA-rhP75 Functional Assay, 2 vials. (93-1009C3)
Catalog No. 502001828
Encompass
The PathHunter U2OS rhTrkA-rhP75 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated rhTrkA and rhP75 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged rhTrkA and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, rhP75 Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the rhesus version of the protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1828 Supplier Eurofins Discoverx Supplier No. 931009C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter U2OS rhTrkA-rhP75 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated rhTrkA and rhP75 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged rhTrkA and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, rhP75 Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the rhesus version of the protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.