Learn More
Eurofins Discoverx PathHunter U2OS SREBP2 Nuclear Translocation Cell Line, 2 vials. (93-0830C3)
Catalog No. 502001381
Encompass
The PathHunter U2OS SREBP2 Nuclear Translocation Cell Line is a stable, engineered cell line for use in studying drug candidates affecting the cholesterol biosynthesis pathway. This cell line enables assessment of ligand (e.g. 5'-UMP) based pathway activation via detection of resulting SREBP2 nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged SREBP2 and nuclear localized EA. Upon pathway activation, induced nuclear translocation of SREBP2 leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.
Catalog No. 50-200-1381 Supplier Eurofins Discoverx Supplier No. 930830C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter U2OS SREBP2 Nuclear Translocation Cell Line is a stable, engineered cell line for use in studying drug candidates affecting the cholesterol biosynthesis pathway. This cell line enables assessment of ligand (e.g. 5'-UMP) based pathway activation via detection of resulting SREBP2 nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged SREBP2 and nuclear localized EA. Upon pathway activation, induced nuclear translocation of SREBP2 leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.