Learn More
Eurofins Discoverx PathHunter U2OS SSTR5 β-Arrestin Cell Line, 2 vials. (93-0402C3)
Catalog No. 502000450
Encompass
The PathHunter SSTR5 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. somatostatin) based activation of SSTR5 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged SSTR5 and EA-tagged β-Arrestin-2. Activation of SSTR5 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0450 Supplier Eurofins Discoverx Supplier No. 930402C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter SSTR5 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. somatostatin) based activation of SSTR5 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged SSTR5 and EA-tagged β-Arrestin-2. Activation of SSTR5 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.