Learn More
Eurofins Discoverx PathHunter U2OS TBXA2R β-Arrestin Cell Line, 2 vials. (93-0322C3)
Catalog No. 502000277
Encompass
The PathHunter TBXA2R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. thromboxane A2, prostaglandins) based activation of TBXA2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged TBXA2R and EA-tagged β-Arrestin-2. Activation of TBXA2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0277 Supplier Eurofins Discoverx Supplier No. 930322C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter TBXA2R cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. thromboxane A2, prostaglandins) based activation of TBXA2R GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged TBXA2R and EA-tagged β-Arrestin-2. Activation of TBXA2R stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.