Learn More
Eurofins Discoverx PathHunter U2OS TGFBR1/ACVR2B Dimerization Cell Line, 2 vials. (93-1046C3)
Supplier:  Eurofins Discoverx 931046C3
Encompass
The PathHunter TGFBR1/ACVR2B Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. GDF-11) induced dimerization of TGFBR1/ACVR2B receptors. This product uses EFC technology. The cell line included overexpresses ACVR2B target protein fused to PK alongside TGFBR1 target protein fused to EA. Upon ligand-induced dimerization of TGFBR1/ACVR2B, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1926
Add to Cart
Request a Quote