Learn More
Eurofins Discoverx PathHunter U2OS TGFBR1/TGFBR2/ENG Dimerization Cell Line, 2 vials. (93-1122C3)
Supplier:  Eurofins Discoverx 931122C3
Encompass
The PathHunter TGFBR1/TGFBR2/ENG Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. TGF-β1) induced dimerization of TGFBR1/TGFBR2/ENG receptors. This product uses EFC technology. The cell line included overexpresses TGFBR1 target protein fused to PK alongside TGFBR2 target protein fused to EA and untagged ENG. Upon ligand-induced dimerization of TGFBR1/TGFBR2/ENG, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-2143
Only null left
Add to Cart
Request a Quote