Learn More
Eurofins Discoverx PathHunter U2OS TIM3/TIM3 Dimerization Cell Line, 2 vials. (93-1082C3)
Catalog No. 502002000
Encompass
The PathHunter TIM3/TIM3 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand induced dimerization of TIM3/TIM3 receptors. This product uses EFC technology. The cell line included overexpresses TIM3 target protein fused to PK alongside TIM3 target protein fused to EA. Upon ligand-induced dimerization of TIM3/TIM3, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.
Catalog No. 50-200-2000 Supplier Eurofins Discoverx Supplier No. 931082C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter TIM3/TIM3 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand induced dimerization of TIM3/TIM3 receptors. This product uses EFC technology. The cell line included overexpresses TIM3 target protein fused to PK alongside TIM3 target protein fused to EA. Upon ligand-induced dimerization of TIM3/TIM3, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/checkpoint.