Learn More
Eurofins Discoverx PathHunter U2OS TORC2 Nuclear Translocation Cell Line, 2 vials. (93-0813C3)
Catalog No. 502001335
Encompass
The PathHunter U2OS TORC2 Nuclear Translocation Cell Line is a stable, engineered cell line for use in studying drug candidates affecting the CRTC/CREB pathway. This cell line enables assessment of ligand (e.g. forskolin) based pathway activation via detection of resulting TORC2 (CRTC2) nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged TORC2 (CRTC2) and nuclear localized EA. Upon pathway activation, induced nuclear translocation of TORC2 leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.
Catalog No. 50-200-1335 Supplier Eurofins Discoverx Supplier No. 930813C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter U2OS TORC2 Nuclear Translocation Cell Line is a stable, engineered cell line for use in studying drug candidates affecting the CRTC/CREB pathway. This cell line enables assessment of ligand (e.g. forskolin) based pathway activation via detection of resulting TORC2 (CRTC2) nuclear translocation. This product uses EFC technology. The included cell line overexpresses ProLabel-tagged TORC2 (CRTC2) and nuclear localized EA. Upon pathway activation, induced nuclear translocation of TORC2 leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.