Learn More
Eurofins Discoverx PathHunter U2OS TSHR (Long Isoform) β-Arrestin-1 Cell Line, 2 vials. (93-0912C3)
Supplier:  Eurofins Discoverx 930912C3
Encompass
The PathHunter TSHR(L) (ARR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. TSH) based activation of TSHR GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged TSHR and EA-tagged β-Arrestin-1. Activation of TSHR stimulates the recruitment of β-Arrestin-1 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1547
Add to Cart
Request a Quote