Learn More
Eurofins Discoverx PathHunter U2OS YES1 Activity Assay, 2 vials. (93-0883C3)
Catalog No. 502001486
Encompass
The PathHunter U2OS YES Activity Assay is a stable, engineered cell line for use in studying small molecule drug candidates targeting the YES kinase domain. The cell line is engineered to express the catalytic domain from YES fused to the cytokine receptor binding domain (CRBD) of JAK2. The cells co-express an Enzyme Acceptor (EA) tagged SH2 domain and a ProLink (PK) tagged Cytokine receptor. The CRBD is designed to bind the CTK catalytic domain to the GHR-PK. Activation of the GHR-PK induces receptor phosphorylation by the YES catalytic domain. This leads to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1486 Supplier Eurofins Discoverx Supplier No. 930883C3
Add to Cart
Add to Cart
Request a Quote
The PathHunter U2OS YES Activity Assay is a stable, engineered cell line for use in studying small molecule drug candidates targeting the YES kinase domain. The cell line is engineered to express the catalytic domain from YES fused to the cytokine receptor binding domain (CRBD) of JAK2. The cells co-express an Enzyme Acceptor (EA) tagged SH2 domain and a ProLink (PK) tagged Cytokine receptor. The CRBD is designed to bind the CTK catalytic domain to the GHR-PK. Activation of the GHR-PK induces receptor phosphorylation by the YES catalytic domain. This leads to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.