Learn More
Enzo Life Sciences U-46619, (1mg), CAS Number: 56985-40-1
SDP
Catalog No. NC9787129
Encompass_Preferred
Prostaglandin H2/thromboxane A2 receptor agonist. Prostaglandin H2/thromboxane A2 receptor agonist; stable analog of thromboxane A2. Induces platelet aggregation at 32nM and activates p38 MAPK. Alternative Name: 9,11-Dideoxy-9alpha,11alpha-methanoepoxy prostaglandin F2alpha, 9,11-Dideoxy-9alpha,11alpha-methanoepoxy prosta5Z,13E-dien-1-oic acid. Formula: C21H34O4. MW: 350.5. CAS Number: 56985-40-1. Purity: ≥98% (HPLC). Appearance: Pale yellow oil. Solubility: Soluble in 100% ethanol, DMSO, dimethyl formamide (100mg/ml) or PBS, pH 7.2 (2mg/ml). Use/Stability: Stable for at least 1 years after receipt when stored at -20°C. We do not recommend storing aqueous solutions for more than one day. Long Term Storage: -20°C
Catalog No. NC9787129 Supplier Enzo Life Sciences Supplier No. BMLPG0230001
Prostaglandin H2/thromboxane A2 receptor agonist. Prostaglandin H2/thromboxane A2 receptor agonist; stable analog of thromboxane A2. Induces platelet aggregation at 32nM and activates p38 MAPK. Alternative Name: 9,11-Dideoxy-9alpha,11alpha-methanoepoxy prostaglandin F2alpha, 9,11-Dideoxy-9alpha,11alpha-methanoepoxy prosta5Z,13E-dien-1-oic acid. Formula: C21H34O4. MW: 350.5. CAS Number: 56985-40-1. Purity: ≥98% (HPLC). Appearance: Pale yellow oil. Solubility: Soluble in 100% ethanol, DMSO, dimethyl formamide (100mg/ml) or PBS, pH 7.2 (2mg/ml). Use/Stability: Stable for at least 1 years after receipt when stored at -20°C. We do not recommend storing aqueous solutions for more than one day. Long Term Storage: -20°C