Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ U5B-ANSI Faceshield Holder for SecureFit™ Safety Helmet

Catalog No. 19071095 Shop All 3M Products
Change view
Click to view available options
Size:
One Size Fits Most
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-071-095 One Size Fits Most
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-071-095 Supplier 3M™ Supplier No. 7100179142
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

U5B-ANSI Faceshield Holder connects a variety of 3M™ faceshields to the 3M™ SecureFit™ Safety Helmet X5000

  • Features a unique system that allows faceshield replacement in seconds.
  • Fits 3M™ SecureFit™ Safety Helmet X5000
  • 3M Catalog# U5B-ANSI

TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Equipment) 3M Faceshields and the 3M SecureFit X5000 Series Safety Helmets
For Use With (Application) Demolition, Forestry, Grinding, Remediation
Material Nylon
Model U5B-ANSI
Product Type Faceshield Holder
Quantity 10/Cs.
Size One Size Fits Most
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.