Learn More
Selleck Chemical LLC U73122 5mg 112648-68-7
SDP
Supplier:  Selleck Chemical LLC S80115MG
Encompass_Preferred
U73122 is a potent phospholipase C (PLC) inhibitor, which reduces agonist-induced Ca2+ increases in platelets and PMN. U73122 potently inhibits human 5-lipoxygenase (5-LO). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5551