Learn More
Medchemexpress LLC U93631 | 152273-12-6 | 99.9% | 299.37 g/mol | C17H21N3O2 | 25 MG
SDP
Catalog No. 5000213705
Encompass_Preferred
A synthetic GABAA receptor ligand intended for in vitro pharmacology and electrophysiology research. Reported activity includes an IC50 of 100 nM and a capacity to induce a rapid, time-dependent decay of GABA-evoked whole-cell chloride currents in recombinant GABAA receptors. The compound is supplied as a high-purity solid and in DMSO stock solutions for laboratory use.
  • Potent GABAA receptor ligand with reported IC50 of 100 nM.
  • Induces rapid, time-dependent decay of GABA-evoked whole-cell Cl-currents.
  • High purity suitable for in vitro studies (≈99.9%).
  • Molecular formula C17H21N3O2 and molecular weight 299.37 g/mol.
  • Available as small solid quantities and DMSO solution stocks for convenient dosing.
Catalog No. 50-002-13705 Supplier Medchemexpress LLC Supplier No. HY10068625MG
Add to Cart
Add to Cart
A synthetic GABAA receptor ligand intended for in vitro pharmacology and electrophysiology research. Reported activity includes an IC50 of 100 nM and a capacity to induce a rapid, time-dependent decay of GABA-evoked whole-cell chloride currents in recombinant GABAA receptors. The compound is supplied as a high-purity solid and in DMSO stock solutions for laboratory use.
  • Potent GABAA receptor ligand with reported IC50 of 100 nM.
  • Induces rapid, time-dependent decay of GABA-evoked whole-cell Cl-currents.
  • High purity suitable for in vitro studies (≈99.9%).
  • Molecular formula C17H21N3O2 and molecular weight 299.37 g/mol.
  • Available as small solid quantities and DMSO solution stocks for convenient dosing.