Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories UA DipTube Controls

Catalog No. 23111377
Change view
Click to view available options
Quantity:
2 x 12 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-377 2 x 12 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-377 Supplier Germaine™ Laboratories Supplier No. 51112
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

UA Diptube Controls are assayed urine controls intended for use in validating the precision of both visual and analyzer reading of urinalysis.

Includes Negative Control (1 x 12 mL) and Positive Control (1 x 12 mL)

Specifications

Content And Storage 2°C to 8°C
Detectable Analytes Ascorbic Acid, Glucose, Bilirubin, Ketones, Specific Gravity, Blood, pH, Protein, Urobilinogen, Nitrite, Leukocytes
For Use With (Equipment) AimStrip™ Urine Reagent Strips and AimStrip™ Urine Analyzer. For use with Urine Reagent Strips.
Type UA Control
Quantity 2 x 12 mL
Stability Stable until the expiration date if stored at 2 to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.