Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories UA Liquid Controls Plus

Catalog No. 23111352
Change view
Click to view available options
Quantity:
2 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-352 2 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-352 Supplier Germaine™ Laboratories Supplier No. 51116
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Germaine™ Laboratories UA Liquid Controls Plus are for use with AimStrip™ Urine Reagent Strips, AimTab Bilirubin Tablets, AimTab Ketone Tablets, and include hCG for pregnancy tests

  • Packaged in dropper bottles for ease of use
  • 2 x 5mL
    • 1 x 5mL Negative
    • 1 x 5mL Positive for glucose, bilirubin, ketone, specific gravity, blood, pH, protein, urobilinogen, nitrite, leukocytes, and hCG

Specifications

Content And Storage 2°C to 8°C
For Use With (Equipment) AimStrip™ Urine Reagent Strips, AimTab Bilirubin Tablets, AimTab Ketone Tablets, and includes hCG for pregnancy tests.
Quantity 2 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.