Learn More
Medchemexpress LLC HY-111434A 25mg , UAA crosslinker 1 (hydrochloride) CAS:1994331-17-7 Purity:>98%
SDP
Catalog No. 502261658
Encompass_Preferred
HY-111434A 25mg UAA crosslinker 1 (hydrochloride) CAS: 1994331-17-7 UAA crosslinker 1 hydrochloride is an amber codon used for non-canonical amino acids (ncAAs) incorporation. The ncAAs can be incorporated into proteins in vivo by making use of the promiscuous activity of certain wildtype and engineered aminoacyl-tRNA synthetases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1658 Supplier Medchemexpress LLC Supplier No. HY111434A25MG
HY-111434A 25mg UAA crosslinker 1 (hydrochloride) CAS: 1994331-17-7 UAA crosslinker 1 hydrochloride is an amber codon used for non-canonical amino acids (ncAAs) incorporation. The ncAAs can be incorporated into proteins in vivo by making use of the promiscuous activity of certain wildtype and engineered aminoacyl-tRNA synthetases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.