Learn More
Apexbio Technology LLC UAMC 00039 dihydrochloride 697797-51-6 50mg
SDP
Catalog No. 5000091425
Encompass_Preferred
UAMC 00039 dihydrochloride (CAS 697797-51-6) is a small molecule inhibitor that selectively targets dipeptidyl peptidase II It acts through a reversible and competitive mechanism exhibiting an IC50 of 0 48 nM In both in vitro and in vivo models UAMC 00039 dihydrochloride has been shown to modulate relevant signaling pathways by suppressing the enzymatic activity of its target This compound is utilized in biomedical research to investigate metabolic disorders and immune system diseases providing a tool for studying the functional role of dipeptidyl peptidase II in various physiological and pathological contexts
Catalog No. 50-000-91425 Supplier Apexbio Technology LLC Supplier No. B578050
Add to Cart
Add to Cart
UAMC 00039 dihydrochloride (CAS 697797-51-6) is a small molecule inhibitor that selectively targets dipeptidyl peptidase II It acts through a reversible and competitive mechanism exhibiting an IC50 of 0 48 nM In both in vitro and in vivo models UAMC 00039 dihydrochloride has been shown to modulate relevant signaling pathways by suppressing the enzymatic activity of its target This compound is utilized in biomedical research to investigate metabolic disorders and immune system diseases providing a tool for studying the functional role of dipeptidyl peptidase II in various physiological and pathological contexts