Learn More
Cayman Chemical UAMC-0039 (hydrochloride), Purity: ≥98%, CAS: 697797-51-6, 10 mg
SDP
Catalog No. 502972663
Encompass_Preferred
A potent inhibitor of DPP-2 (IC50 = 0.48 nM); inhibits DPP-4 (IC50 = 165 mM)
Catalog No. 50-297-2663 Supplier Cayman Chemical Supplier No. 1534010 MG
A potent inhibitor of DPP-2 (IC50 = 0.48 nM); inhibits DPP-4 (IC50 = 165 mM)