Learn More
Medchemexpress LLC HY-112909A 10mg Medchemexpress, UAMC-3203 hydrochloride CAS:2271358-65-5 Purity:>98%
SDP
Catalog No. 501969203
Encompass_Preferred
Medchemexpress, HY-112909A 10mg UAMC-3203 HCl CAS:2271358-65-5 UAMC-3203 hydrochloride is a potent and selective Ferroptosis inhibitor with an IC50 of 12 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9203 Supplier Medchemexpress LLC Supplier No. HY112909A10MG
Medchemexpress, HY-112909A 10mg UAMC-3203 HCl CAS:2271358-65-5 UAMC-3203 hydrochloride is a potent and selective Ferroptosis inhibitor with an IC50 of 12 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.