Learn More
Medchemexpress LLC UAMC-3203 hydrochloride | 2271358-65-5 | 98.1% | 508.12 | 100 MG
SDP
Catalog No. 5000439826
Encompass_Preferred
UAMC-3203 hydrochloride is a potent and selective ferroptosis inhibitor with an IC50 of 12 nM. It is for research use only and not sold to patients.
  • No toxicity observed in mice after repeated injections.
  • Available in solid and solution forms.
  • Ships at room temperature in continental US.
  • Store at 4°C, sealed, away from moisture and light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light.
Catalog No. 50-004-39826 Supplier Medchemexpress LLC Supplier No. HY112909A100MG
UAMC-3203 hydrochloride is a potent and selective ferroptosis inhibitor with an IC50 of 12 nM. It is for research use only and not sold to patients.
  • No toxicity observed in mice after repeated injections.
  • Available in solid and solution forms.
  • Ships at room temperature in continental US.
  • Store at 4°C, sealed, away from moisture and light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture and light.