Learn More
Medchemexpress LLC UAMC-3203 hydrochloride | 2271358-65-5 | 98.1% | 508.12 | 50 MG
SDP
Catalog No. 5000438779
Encompass_Preferred
UAMC-3203 hydrochloride is a potent and selective Ferroptosis inhibitor with an IC50 of 12 nM. It is for research use only and not sold to patients. No toxicity was observed in mice after repeated intraperitoneal injections (20 μmol/kg daily, over 4 weeks).
  • Potent and selective Ferroptosis inhibitor.
  • IC50 of 12 nM for Ferroptosis.
  • Inhibits erastin-induced ferroptosis in human IMR32 cells with an IC50 of 10 nM.
  • No observed toxicity in mice at 20 μmol/kg over 4 weeks.
Catalog No. 50-004-38779 Supplier Medchemexpress LLC Supplier No. HY112909A50MG
UAMC-3203 hydrochloride is a potent and selective Ferroptosis inhibitor with an IC50 of 12 nM. It is for research use only and not sold to patients. No toxicity was observed in mice after repeated intraperitoneal injections (20 μmol/kg daily, over 4 weeks).
  • Potent and selective Ferroptosis inhibitor.
  • IC50 of 12 nM for Ferroptosis.
  • Inhibits erastin-induced ferroptosis in human IMR32 cells with an IC50 of 10 nM.
  • No observed toxicity in mice at 20 μmol/kg over 4 weeks.