Learn More
Cayman Chemical UAMC-3203 (hydrochloride), Purity: ≥98%, CAS: 2271358-65-5, 10 mg
SDP
Catalog No. 502990561
Encompass_Preferred
A ferroptosis inhibitor (IC50 = 10 nM for erastin-induced ferroptosis in IMR-32 cells); decreases iron-induced plasma LDH levels in a mouse model of acute iron poisoning at 20 µmol/kg; not toxic to mice at 20 µmol/kg for four weeks
Catalog No. 50-299-0561 Supplier Cayman Chemical Supplier No. 2934610 MG
A ferroptosis inhibitor (IC50 = 10 nM for erastin-induced ferroptosis in IMR-32 cells); decreases iron-induced plasma LDH levels in a mouse model of acute iron poisoning at 20 µmol/kg; not toxic to mice at 20 µmol/kg for four weeks