Learn More
Medchemexpress LLC HY-101769 5mg Medchemexpress, UAMC00039 (dihydrochloride) CAS:697797-51-6 Purity:>98%
SDP
Catalog No. 501932209
Encompass_Preferred
Medchemexpress, HY-101769 5mg UAMC00039 (dihydrochloride) CAS:697797-51-6 UAMC00039 dihydrochloride is a potent, reversible and competitive dipeptidyl peptidase II inhibitor with an IC 50 of 0.48 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2209 Supplier Medchemexpress LLC Supplier No. HY1017695MG
Medchemexpress, HY-101769 5mg UAMC00039 (dihydrochloride) CAS:697797-51-6 UAMC00039 dihydrochloride is a potent, reversible and competitive dipeptidyl peptidase II inhibitor with an IC 50 of 0.48 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.