Learn More
Medchemexpress LLC UAMC00039 dihydrochloride | 697797-51-6 | 99.5% | 382.76 | C16H26Cl3N3O | 2 MG
SDP
Catalog No. 5000245706
Encompass_Preferred
UAMC00039 dihydrochloride is a potent, reversible, competitive inhibitor of dipeptidyl peptidase II (DPP-II) with a reported IC50 of 0.48 nM. It is supplied as the dihydrochloride salt and intended as a high-purity research reagent for biochemical assays, selectivity profiling, and mechanism-of-action studies.
  • High potency: IC50 0.48 nM against DPP-II.
  • High purity suitable for biochemical and pharmacological assays (≈99.5%).
  • Provided as the dihydrochloride salt to improve stability and solubility.
  • Available in small milligram quantities and solution formats for screening.
  • Suitable for enzyme inhibition, selectivity profiling, and preclinical research applications.
Catalog No. 50-002-45706 Supplier Medchemexpress LLC Supplier No. HY1017692MG
UAMC00039 dihydrochloride is a potent, reversible, competitive inhibitor of dipeptidyl peptidase II (DPP-II) with a reported IC50 of 0.48 nM. It is supplied as the dihydrochloride salt and intended as a high-purity research reagent for biochemical assays, selectivity profiling, and mechanism-of-action studies.
  • High potency: IC50 0.48 nM against DPP-II.
  • High purity suitable for biochemical and pharmacological assays (≈99.5%).
  • Provided as the dihydrochloride salt to improve stability and solubility.
  • Available in small milligram quantities and solution formats for screening.
  • Suitable for enzyme inhibition, selectivity profiling, and preclinical research applications.