Learn More
Aviva Systems Biology Corp Uap1 Antibody - C-terminal region (ARP49212_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49212_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Uap1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AA420407, AA437972, AGX-1, AGX-2, AGX1, AgX, ESTM38, SPAG2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-405