Learn More
Aviva Systems Biology Corp Uap1l1 Antibody - middle region (ARP49211_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49211_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Uap1l1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 5730445F03RikApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-404