Learn More
Electron Microscopy Sciences UAR-EMS Uranyl Acetate Replacement 25 ML
SDP
Catalog No. NC1222619
Encompass_Preferred
A non-radioactive substitute for uranyl acetate with comparable results. Avoids the tedious authorization, shipping and disposal issues involving uranyl acetate without sacrificing performance. Two lanthanide salts, samarium and gadolinium triacetate, effectively used for staining plastic-embedded animal and plant tissues. Also for negative staining of macromolecules. Not recommended as a fixative.
Catalog No. NC1222619 Supplier Electron Microscopy Sciences Supplier No. 22405
A non-radioactive substitute for uranyl acetate with comparable results. Avoids the tedious authorization, shipping and disposal issues involving uranyl acetate without sacrificing performance. Two lanthanide salts, samarium and gadolinium triacetate, effectively used for staining plastic-embedded animal and plant tissues. Also for negative staining of macromolecules. Not recommended as a fixative.