Learn More
Perkin Elmer US LLC UATR Sample Position Plate - 7 mm
Catalog No. 502113395
Encompass_Preferred
The PerkinElmer Universal ATR (UATR) Accessory is ideal for the general IR analysis of solids and liquids. The UATR comes in a variety of crystal configurations as well as single/multiple reflections capabilities. It produces high quality spectra through the use of a pressure arm. The pressure arm force indicator ensures sample-to-sample and operator-to-operator reproducibility. The accessory is recognized when it is placed in the spectrometer's sample compartment, the optics are automatically optimized, and the system is ready to be used without additional alignment. This UATR is compatible with the Spectrum One/100/400/65 and Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-3395 Supplier Perkin Elmer US LLC Supplier No. L1202024
The PerkinElmer Universal ATR (UATR) Accessory is ideal for the general IR analysis of solids and liquids. The UATR comes in a variety of crystal configurations as well as single/multiple reflections capabilities. It produces high quality spectra through the use of a pressure arm. The pressure arm force indicator ensures sample-to-sample and operator-to-operator reproducibility. The accessory is recognized when it is placed in the spectrometer's sample compartment, the optics are automatically optimized, and the system is ready to be used without additional alignment. This UATR is compatible with the Spectrum One/100/400/65 and Spectrum 3/Frontier spectrometers.