Learn More
Perkin Elmer US LLC UATR Top Plate - ZnSe, 9-Reflection
Catalog No. 502113383
Encompass_Preferred
This plate is compatible with the UATR accessories that are compatible with the Spectrum One/100/400/65 and Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-3383 Supplier Perkin Elmer US LLC Supplier No. L1200366
Add to Cart
Add to Cart
Request a Quote
This plate is compatible with the UATR accessories that are compatible with the Spectrum One/100/400/65 and Spectrum 3/Frontier spectrometers.