Learn More
Ubpbio UB ALDEHYDE, 50UG
Catalog No. 501126927
Encompass_Preferred
Ubiquitin Derivatives – Ubiquitin Aldehyde (UBPBio catalog# F3100) – Ub aldehyde is a potent and highly specific inhibitor of cysteine deubiquitinating enzymes including the Ub C-terminal hydrolases (UCHs), the Ub-specific proteases (USPs), the ovarian tumor domains (OTUs) proteases and the Machado-Josephine domain (MJD) proteases. Typical working concentration is 2-5 µM in vitro.
Catalog No. 50-112-6927 Supplier Ubpbio Supplier No. F3100
Ubiquitin Derivatives – Ubiquitin Aldehyde (UBPBio catalog# F3100) – Ub aldehyde is a potent and highly specific inhibitor of cysteine deubiquitinating enzymes including the Ub C-terminal hydrolases (UCHs), the Ub-specific proteases (USPs), the ovarian tumor domains (OTUs) proteases and the Machado-Josephine domain (MJD) proteases. Typical working concentration is 2-5 µM in vitro.