Learn More
Enzo Life Sciences Mono- and polyubiquitinylated conjugates recombinant monoclonal antibody (UBCJ2) (100 µg)
SDP
Supplier:  Enzo Life Sciences ENZABS8400100
Encompass_Preferred
Clone: UBCJ2. Host: Mouse. Isotype: IgG1κ. Immunogen: Poly-ubiquitinylated lysozyme. UniProt ID: P0CG47 (UBB), P0CG48 (UBC). Source: Purified from tissue culture supernatant. Species reactivity: Species independent. Applications: ELISA, Flow Cytometry, ICC, WB. Purification: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-210-0453