Learn More
Enzo Life Sciences Ubiquitin activating enzyme (CT) polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW83950025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 1040-1058 of C-terminal human Ub E1 (ubiquitin E1 activating enzyme). Source: Purified from rabbit serum. Species reactivity: Species independent. Specificity: Recognizes Ub E1 in a wide range of species. Applications: WB. Formulation: Liquid. In PBS containing 0.01% sodium azide.
Catalog No. 50-201-1635