Learn More
LifeSensors ITCH 25µg
SDP
Supplier:  LifeSensors UB03150025
Encompass_Preferred
ITCH is a HECT E3, which influences immunity, the skin, and plays a role in cancer by targeting various transcription factors and growth factors. Used in neurodegeneration research and protein regulation studies, enabling targeted ubiquitination analysis.
Catalog No. NC2388853