Learn More
LifeSensors Parkin (Non-phosphorylated)
SDP
Catalog No. NC3351368
Encompass_Preferred
Encoded by the PARK2 gene, the E3 ligase Parkin is part of the multi-protein E3 complex that encodes substrate proteins for degradation in the Ubiquitin-Proteasome Pathway. The precise function of this protein is unknown; however, mutations in this gene are known to cause a familial form of Parkinson’s disease known as autosomal recessive juvenile Parkinson’s disease (AR-JP). Parkin is described to be necessary for mitophagy (autophagy of mitochondria).
Catalog No. NC3351368 Supplier LifeSensors Supplier No. UB03170025
Add to Cart
Add to Cart
Encoded by the PARK2 gene, the E3 ligase Parkin is part of the multi-protein E3 complex that encodes substrate proteins for degradation in the Ubiquitin-Proteasome Pathway. The precise function of this protein is unknown; however, mutations in this gene are known to cause a familial form of Parkinson’s disease known as autosomal recessive juvenile Parkinson’s disease (AR-JP). Parkin is described to be necessary for mitophagy (autophagy of mitochondria).