Learn More
Sino Biological Recombinant Human MGRN1 Protein (GST Tag), Active
Catalog No. 502574337
Encompass_Preferred
Recombinant human MGRN1 (2-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4337 Supplier Sino Biological Supplier No. M287380G100
Recombinant human MGRN1 (2-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.