Learn More
Sino Biological Recombinant Human Ubiquitin Activating Enzyme E1/UBA1 Protein (Full Length, GST Tag), Active
Catalog No. 502567570
Encompass_Preferred
Recombinant full length human UBA1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-256-7570 Supplier Sino Biological Supplier No. U201380G20
Recombinant full length human UBA1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.