Learn More
Aviva Systems Biology Corp UBA3 Antibody - N-terminal region (ARP50493_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50493_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human UBA3. Alternate names: DKFZp566J164, MGC22384, UBE1C, hUBA3ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60357