Learn More
Aviva Systems Biology Corp Uba5 Antibody - N-terminal region (ARP49879_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49879_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Uba5. It was validated on Western Blot. . Alternate names: 5730525G14Rik, AW240750, Ube1dc1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-461