Learn More
Aviva Systems Biology Corp UBA6 Antibody - middle region (ARP59384_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59384_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBA6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human UBA6. Alternate names: FLJ10808, FLJ23367, MOP-4, UBE1L2, E1-L2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-678