Learn More
Enzo Life Sciences UBA6 (human) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences BMLPW05250100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 1034-1052 of human UBA6 (UbE1L2). Source: From rabbit serum. Species reactivity: Human. Applications: WB. Formulation: Liquid. Antiserum containing 0.01% sodium azide.
Catalog No. 50-201-1534