Learn More
Aviva Systems Biology Corp UBAC2 Antibody - middle region (ARP55756_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55756_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBAC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UBAC2. Alternate names: FLJ26351, FLJ30001, FLJ30548, FLJ42413, MGC90487, PHGDHL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61600