Learn More
Aviva Systems Biology Corp UBASH3A Antibody - middle region (ARP45769_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45769_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBASH3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UBASH3A. Alternate names: CLIP4, STS-2, TULA, TULA-1ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-59133