Learn More
Canon Instrument Co Ubbelohde Viscometer, Size 0, Calibrated
SDP
Catalog No. NC1744379
Encompass_Preferred
The Ubbelohde Viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104. Minimum sample volume is 11 mL. Minimum bath depth is 241 mm (9.5 in). Unity factor series has a constant within +/- 0.2% of the values listed. Approximate Constant (mm²/s²): 0.001. Viscosity Range (mm²/s): 0.3-1.0
Catalog No. NC1744379 Supplier Canon Instrument Co Supplier No. 9721R50
The Ubbelohde Viscometer measures kinematic viscosity of transparent Newtonian liquids according to ASTM D445 and ISO 3104. Minimum sample volume is 11 mL. Minimum bath depth is 241 mm (9.5 in). Unity factor series has a constant within +/- 0.2% of the values listed. Approximate Constant (mm²/s²): 0.001. Viscosity Range (mm²/s): 0.3-1.0