Learn More
Enzo Life Sciences UbcH1 (human), (recombinant) (His-tag) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW90200100
Encompass_Preferred
Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 20mM TRIS-HCl, pH 7.5, containing 0.5mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2295